Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119986.3 | NP_195537.2 | 13 - 14 | 967 | 99 | |
NM_179186.1 | NP_849517.1 | 13 - 14 | 967 | 99 | ||
GenBank mRNA | AJ243198.1 | - | 13 - 14 | 881 | 99 | |
AY099838.1 | - | 13 - 14 | 938 | 99 | ||
BT000334.1 | - | 13 - 14 | 851 | 99 | ||
KJ138715.1 | - | 13 - 14 | 851 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |