Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179215.2 | NP_849546.1 | 1 | 700 | 83 | |
GenBank mRNA | AY087095.1 | - | 1 | 654 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |