Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nPAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036701.1 | NP_001031778.1 | 1 - 2 | 490 | 83 | |
NM_001036702.1 | NP_001031779.1 | 1 - 2 | 498 | 83 | ||
NM_001125630.1 | NP_001119102.1 | 1 - 2 | 490 | 83 | ||
NM_001125631.2 | NP_001119103.1 | 1 - 2 | 559 | 83 | ||
NM_001342179.1 | NP_001329665.1 | 1 - 2 | 559 | 83 | ||
NM_001342180.1 | NP_001329664.1 | 1 - 2 | 559 | 83 | ||
NM_001342181.1 | NP_001329663.1 | 1 - 2 | 559 | 83 | ||
NM_001342182.1 | NP_001329669.1 | 1 - 2 | 559 | 83 | ||
NM_001342183.1 | NP_001329668.1 | 1 - 2 | 549 | 83 | ||
NM_001342184.1 | NP_001329667.1 | 1 - 2 | 549 | 83 | ||
NM_001342185.1 | NP_001329666.1 | 1 - 2 | 525 | 83 | ||
NM_119438.4 | NP_195011.4 | 1 - 2 | 490 | 83 | ||
NM_179230.2 | NP_849561.1 | 1 - 2 | 559 | 83 | ||
NM_179231.2 | NP_849562.2 | 1 - 2 | 490 | 83 | ||
NM_202939.3 | NP_974668.1 | 1 - 2 | 559 | 83 | ||
NM_202940.3 | NP_974669.1 | 1 - 2 | 559 | 83 | ||
NM_202941.3 | NP_974670.2 | 1 - 2 | 559 | 83 | ||
GenBank mRNA | AK316738.1 | - | 1 - 2 | 491 | 83 | |
AK318704.1 | - | 1 - 2 | 495 | 83 | ||
AY069880.1 | - | 1 - 2 | 490 | 83 | ||
BT000485.1 | - | 1 - 2 | 136 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |