Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035860.1 | NP_001030937.1 | 2 - 3 | 244 | 92 | |
NM_001331368.1 | NP_001320441.1 | 2 - 3 | 264 | 92 | ||
NM_100176.4 | NP_563670.1 | 2 - 3 | 246 | 92 | ||
NM_179250.1 | NP_849581.1 | 2 - 3 | 251 | 92 | ||
GenBank mRNA | AF320055.1 | - | 2 - 3 | 213 | 92 | |
AK228359.1 | - | 2 - 3 | 251 | 92 | ||
BT005712.1 | - | 2 - 3 | 251 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |