Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100191.4 | NP_563674.1 | 8 - 9 | 1189 | 63 | |
NM_179252.3 | NP_849583.1 | 9 - 10 | 1383 | 63 | ||
GenBank mRNA | AY070723.1 | - | 9 - 10 | 1240 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |