Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100201.3 | NP_171818.1 | 5 - 6 | 1435 | 76 | |
NM_179253.1 | NP_849584.1 | 4 - 5 | 1498 | 76 | ||
GenBank mRNA | AY062471.1 | - | 5 - 6 | 1435 | 76 | |
AY090788.1 | - | 4 - 5 | 1498 | 76 | ||
AY090915.1 | - | 4 - 5 | 1298 | 76 | ||
BT010344.1 | - | 4 - 5 | 1298 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |