Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLX2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100494.5 | NP_563760.1 | 4 - 5 | 865 | 77 | |
NM_179268.2 | NP_849599.1 | 4 - 5 | 622 | 77 | ||
GenBank mRNA | AK227097.1 | - | 4 - 5 | 579 | 77 | |
AY085754.1 | - | 4 - 5 | 639 | 77 | ||
AY099584.1 | - | 4 - 5 | 572 | 77 | ||
AY128833.1 | - | 4 - 5 | 564 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |