Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331755.1 | NP_001322007.1 | 14 - 15 | 5587 | 83 | |
NM_100680.4 | NP_563806.1 | 14 - 15 | 5584 | 83 | ||
NM_179277.1 | NP_849608.1 | 13 - 14 | 5316 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |