Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331755.1 | NP_001322007.1 | 7 - 8 | 3228 | 111 | |
NM_100680.4 | NP_563806.1 | 7 - 8 | 3225 | 111 | ||
NM_179277.1 | NP_849608.1 | 6 - 7 | 2957 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |