Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035934.2 | NP_001031011.1 | 3 - 4 | 506 | 89 | |
NM_100797.6 | NP_172398.1 | 3 - 4 | 593 | 89 | ||
NM_179292.3 | NP_849623.1 | 3 - 4 | 593 | 89 | ||
GenBank mRNA | AK316689.1 | - | 3 - 4 | 490 | 89 | |
AY090944.1 | - | 3 - 4 | 516 | 89 | ||
AY117313.1 | - | 3 - 4 | 418 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |