Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331836.1 | NP_001318964.1 | 3 - 4 | 492 | 82 | |
NM_001331837.1 | NP_001318965.1 | 3 - 4 | 552 | 82 | ||
NM_001331838.1 | NP_001322700.1 | 2 - 3 | 916 | 82 | ||
NM_001331839.1 | NP_001322701.1 | 1 - 2 | 889 | 82 | ||
NM_100824.3 | NP_172424.1 | 2 - 3 | 465 | 82 | ||
NM_179295.3 | NP_849626.1 | 2 - 3 | 525 | 82 | ||
GenBank mRNA | AF100166.1 | - | 2 - 3 | 157 | 82 | |
AF251693.1 | - | 1 - 2 | 74 | 82 | ||
AK117255.1 | - | 2 - 3 | 210 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |