Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035948.1 | NP_001031025.1 | 6 - 7 | 640 | 102 | |
NM_001035949.3 | NP_001031026.1 | 6 - 7 | 861 | 102 | ||
NM_001198039.1 | NP_001184968.1 | 6 - 7 | 773 | 102 | ||
NM_101055.4 | NP_172648.1 | 6 - 7 | 897 | 102 | ||
NM_179312.4 | NP_849643.1 | 6 - 7 | 872 | 102 | ||
NM_179313.2 | NP_849644.2 | 6 - 7 | 617 | 102 | ||
GenBank mRNA | AB050553.1 | - | 6 - 7 | 618 | 102 | |
AF428272.1 | - | 6 - 7 | 618 | 102 | ||
AK316841.1 | - | 6 - 7 | 619 | 102 | ||
AK318730.2 | - | 6 - 7 | 613 | 102 | ||
AY074569.1 | - | 6 - 7 | 618 | 102 | ||
AY087874.1 | - | 6 - 7 | 653 | 102 | ||
AY097360.1 | - | 5 - 6 | 555 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |