Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332005.1 | NP_001321932.1 | 8 - 9 | 1269 | 90 | |
NM_001332006.1 | NP_001321931.1 | 8 - 9 | 1269 | 90 | ||
NM_001332007.1 | NP_001321930.1 | 6 - 7 | 879 | 90 | ||
NM_101058.2 | NP_172651.1 | 8 - 9 | 1214 | 90 | ||
NM_179316.3 | NP_849647.1 | 8 - 9 | 1309 | 90 | ||
GenBank mRNA | AY091115.1 | - | 8 - 9 | 1221 | 90 | |
AY122978.1 | - | 8 - 9 | 1171 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |