Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332851.1 | NP_001320501.1 | 4 - 5 | 1243 | 120 | |
NM_102680.2 | NP_174233.2 | 4 - 5 | 851 | 120 | ||
NM_179396.3 | NP_849727.1 | 4 - 5 | 1000 | 120 | ||
GenBank mRNA | AY062613.1 | - | 4 - 5 | 887 | 120 | |
AY070368.1 | - | 4 - 5 | 890 | 120 | ||
AY091327.1 | - | 3 - 4 | 727 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |