Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179456.3 | NP_849787.1 | 3 - 4 | 496 | 106 | |
GenBank mRNA | AB085617.1 | - | 2 - 3 | 122 | 106 | |
AK317094.1 | - | 3 - 4 | 329 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |