Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103882.4 | NP_564559.1 | 2 - 3 | 366 | 69 | |
NM_179458.3 | NP_849789.1 | 2 - 3 | 379 | 69 | ||
NM_202269.2 | NP_973998.1 | 2 - 3 | 455 | 69 | ||
GenBank mRNA | AY062736.1 | - | 2 - 3 | 291 | 69 | |
AY519539.1 | - | 1 - 2 | 152 | 69 | ||
BT009677.1 | - | 1 - 2 | 152 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |