Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112103.3 | NP_187874.2 | 4 - 5 | 903 | 107 | |
GenBank mRNA | AY024357.1 | - | 4 - 5 | 903 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |