Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112111.3 | NP_187881.1 | 1 - 2 | 803 | 59 | |
GenBank mRNA | AF033535.1 | - | 1 - 2 | 799 | 59 | |
AK226531.1 | - | 1 - 2 | 806 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |