Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338008.1 | NP_001326839.1 | 1 - 2 | 696 | 87 | |
NM_112117.5 | NP_187887.3 | 1 - 2 | 696 | 87 | ||
GenBank mRNA | AY279398.1 | - | 1 - 2 | 159 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |