Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112163.6 | NP_187929.4 | 7 - 8 | 1148 | 68 | |
GenBank mRNA | AY063883.1 | - | 7 - 8 | 1107 | 68 | |
AY142588.1 | - | 7 - 8 | 1076 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |