Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112191.3 | NP_187954.1 | 1 - 2 | 312 | 136 | |
GenBank mRNA | AF145452.1 | - | 1 - 2 | 249 | 136 | |
AK229269.1 | - | 1 - 2 | 310 | 136 | ||
BT024889.1 | - | 1 - 2 | 171 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |