Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112210.3 | NP_187973.1 | 7 - 8 | 1000 | 66 | |
GenBank mRNA | BT003947.1 | - | 7 - 8 | 884 | 66 | |
BT005042.1 | - | 6 - 7 | 794 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |