Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112210.3 | NP_187973.1 | 9 - 10 | 1390 | 60 | |
GenBank mRNA | BT003947.1 | - | 9 - 10 | 1274 | 60 | |
BT005042.1 | - | 8 - 9 | 1184 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |