Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWINV5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338076.1 | NP_001326013.1 | 3 - 4 | 1090 | 106 | |
NM_001338077.1 | NP_001326012.1 | 2 - 3 | 904 | 106 | ||
NM_001338078.1 | NP_001326014.1 | 2 - 3 | 1046 | 106 | ||
NM_112231.2 | NP_187994.1 | 2 - 3 | 1065 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |