Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATBRXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112254.3 | NP_188016.2 | 2 - 3 | 576 | 74 | |
NM_202580.2 | NP_974309.1 | 3 - 4 | 1149 | 74 | ||
GenBank mRNA | AK117522.1 | - | 2 - 3 | 573 | 74 | |
BT029002.1 | - | 2 - 3 | 515 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |