Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084681.1 | NP_001078150.1 | 11 - 12 | 1676 | 78 | |
NM_001084682.1 | NP_001078151.1 | 11 - 12 | 1653 | 78 | ||
NM_001202957.1 | NP_001189886.1 | 11 - 12 | 1787 | 78 | ||
NM_001338097.1 | NP_001325470.1 | 11 - 12 | 2200 | 78 | ||
NM_112255.4 | NP_566471.1 | 11 - 12 | 1984 | 78 | ||
GenBank mRNA | AY045922.1 | - | 11 - 12 | 1680 | 78 | |
AY079365.1 | - | 10 - 11 | 1414 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |