Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202959.1 | NP_001189888.1 | 3 - 4 | 514 | 83 | |
NM_112267.4 | NP_566478.1 | 3 - 4 | 488 | 83 | ||
NM_202581.2 | NP_974310.1 | 2 - 3 | 611 | 83 | ||
GenBank mRNA | AF446890.1 | - | 3 - 4 | 424 | 83 | |
AY052677.1 | - | 3 - 4 | 467 | 83 | ||
AY087529.1 | - | 3 - 4 | 473 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |