Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338114.1 | NP_001326872.1 | 2 - 3 | 408 | 65 | |
NM_112278.3 | NP_188037.1 | 1 - 2 | 750 | 65 | ||
GenBank mRNA | AK221834.1 | - | 1 - 2 | 326 | 65 | |
AY054230.1 | - | 1 - 2 | 304 | 65 | ||
AY062690.1 | - | 1 - 2 | 306 | 65 | ||
BT002522.1 | - | 1 - 2 | 306 | 65 | ||
BT008484.1 | - | 1 - 2 | 263 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |