Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112280.1 | NP_188039.1 | 2 - 3 | 409 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |