Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 31 - 32 | 3650 | 78 | |
NM_001338124.1 | NP_001326811.1 | 30 - 31 | 3800 | 78 | ||
NM_112317.3 | NP_188075.2 | 30 - 31 | 3887 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |