Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112323.5 | NP_188080.1 | 1 - 2 | 480 | 74 | |
GenBank mRNA | AY052208.1 | - | 1 - 2 | 344 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |