Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112323.5 | NP_188080.1 | 2 - 3 | 689 | 89 | |
GenBank mRNA | AY052208.1 | - | 2 - 3 | 553 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |