Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338141.1 | NP_001326778.1 | 7 - 8 | 1423 | 96 | |
NM_112356.4 | NP_188112.1 | 8 - 9 | 1449 | 96 | ||
GenBank mRNA | AF071788.1 | - | 8 - 9 | 1303 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |