Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112390.5 | NP_188145.1 | 1 - 2 | 1389 | 62 | |
GenBank mRNA | AJ011610.1 | - | 1 - 2 | 402 | 62 | |
AK118611.1 | - | 1 - 2 | 449 | 62 | ||
BT003714.1 | - | 1 - 2 | 322 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |