Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125161.1 | NP_001118633.1 | 2 - 3 | 792 | 113 | |
NM_001338153.1 | NP_001326301.1 | 3 - 4 | 998 | 113 | ||
NM_112397.4 | NP_188152.3 | 4 - 5 | 970 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |