Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119316.5 | NP_194895.1 | 6 - 7 | 1002 | 95 | |
GenBank mRNA | AY142532.1 | - | 6 - 7 | 895 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |