Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119343.4 | NP_194920.1 | 4 - 5 | 970 | 120 | |
GenBank mRNA | AJ005195.1 | - | 4 - 5 | 780 | 120 | |
AK228149.1 | - | 4 - 5 | 851 | 120 | ||
BT003158.1 | - | 4 - 5 | 694 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |