Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342124.1 | NP_001329752.1 | 4 - 5 | 668 | 96 | |
NM_119353.4 | NP_194929.2 | 6 - 7 | 1016 | 96 | ||
GenBank mRNA | AB206554.1 | - | 2 - 3 | 202 | 96 | |
AK228013.1 | - | 6 - 7 | 871 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |