Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342129.1 | NP_001329706.1 | 11 - 12 | 1655 | 72 | |
NM_119372.3 | NP_194947.2 | 11 - 12 | 1697 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |