Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342129.1 | NP_001329706.1 | 5 - 6 | 606 | 70 | |
NM_119372.3 | NP_194947.2 | 5 - 6 | 606 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |