Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342130.1 | NP_001329438.1 | 2 - 3 | 615 | 100 | |
NM_119380.3 | NP_567891.1 | 2 - 3 | 741 | 100 | ||
GenBank mRNA | AB493715.1 | - | 2 - 3 | 594 | 100 | |
AB493716.1 | - | 2 - 3 | 594 | 100 | ||
AK229689.1 | - | 2 - 3 | 686 | 100 | ||
AY669800.1 | - | 2 - 3 | 594 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |