Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119393.3 | NP_194967.1 | 6 - 7 | 1475 | 84 | |
GenBank mRNA | AK226243.1 | - | 6 - 7 | 1154 | 84 | |
BT008654.1 | - | 6 - 7 | 1163 | 84 | ||
KJ138727.1 | - | 6 - 7 | 1019 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |