Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125626.1 | NP_001119098.1 | 8 - 9 | 1124 | 69 | |
NM_119404.3 | NP_567895.1 | 8 - 9 | 1337 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |