Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIFY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119409.4 | NP_567898.1 | 5 - 6 | 1163 | 62 | |
GenBank mRNA | BT006431.1 | - | 5 - 6 | 998 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |