Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036696.1 | NP_001031773.1 | 1 - 2 | 304 | 82 | |
NM_119417.4 | NP_194991.1 | 1 - 2 | 506 | 82 | ||
NM_202936.1 | NP_974665.1 | 1 - 2 | 313 | 82 | ||
GenBank mRNA | AY052233.1 | - | 1 - 2 | 304 | 82 | |
BT003093.1 | - | 1 - 2 | 256 | 82 | ||
U81239.1 | - | 1 - 2 | 261 | 82 | ||
Z83202.1 | - | 302 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |