Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119421.5 | NP_567901.1 | 1 - 2 | 507 | 122 | |
GenBank mRNA | AK175236.1 | - | 1 - 2 | 178 | 122 | |
AK176859.1 | - | 1 - 2 | 181 | 122 | ||
AK220670.1 | - | 1 - 2 | 181 | 122 | ||
AK229442.1 | - | 1 - 2 | 178 | 122 | ||
BT024768.1 | - | 1 - 2 | 133 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |