Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119421.5 | NP_567901.1 | 3 - 4 | 788 | 82 | |
GenBank mRNA | AK175236.1 | - | 3 - 4 | 459 | 82 | |
AK176859.1 | - | 3 - 4 | 462 | 82 | ||
AK220670.1 | - | 3 - 4 | 462 | 82 | ||
AK229442.1 | - | 3 - 4 | 459 | 82 | ||
BT024768.1 | - | 3 - 4 | 414 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |