Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119437.3 | NP_195010.1 | 3 - 4 | 437 | 119 | |
GenBank mRNA | AK227471.1 | - | 3 - 4 | 326 | 119 | |
BT006001.1 | - | 3 - 4 | 190 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |