Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342189.1 | NP_001328067.1 | 2 - 3 | 1701 | 134 | |
NM_119452.4 | NP_195024.1 | 3 - 4 | 1659 | 134 | ||
GenBank mRNA | AY072174.1 | - | 3 - 4 | 1554 | 134 | |
AY096513.1 | - | 1 - 2 | 847 | 134 | ||
X80126.1 | - | 1150 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |