Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125635.2 | NP_001119107.1 | 5 - 6 | 870 | 120 | |
NM_001342196.1 | NP_001328068.1 | 5 - 6 | 916 | 120 | ||
NM_001342197.1 | NP_001328069.1 | 5 - 6 | 870 | 120 | ||
NM_119459.4 | NP_195031.2 | 4 - 5 | 984 | 120 | ||
NM_202943.3 | NP_974672.1 | 4 - 5 | 984 | 120 | ||
NM_202944.3 | NP_974673.2 | 5 - 6 | 870 | 120 | ||
GenBank mRNA | AY062551.1 | - | 4 - 5 | 981 | 120 | |
BT008453.1 | - | 3 - 4 | 790 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |